Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Proteasome Substrate III, Fluorogenic
GSA_VA

Catalog No. 5391425MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-914-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-914-25MG Supplier MilliporeSigma™ Supplier No. 5391425MG
Add to Cart
Add to Cart

Proteasome Substrate III, Fluorogenic, is a substrate for proteasome. Also acts as a substrate for calpain, chymotrypsin, and ingensin.

Peptide Sequence: Suc-Leu-Leu-Val-Tyr-AMC

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Proteasome
Molecular Formula C40H53N5O10
Formula Weight 763.9g/mol
Solubility DMSO
Content And Storage Protect from light
Form Amorphous Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.