Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Proteasome Substrate VI, Fluorogenic

Catalog No. 5391495MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-914-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-914-95MG Supplier MilliporeSigma™ Supplier No. 5391495MG
Add to Cart
Add to Cart

A fluorogenic substrate probe useful for assaying trypsin-like activity of proteasome.

  • Packaged Under Inert Gas
  • Peptide Sequence: Z-Ala-Arg-Arg-AMC
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Formulation Supplied as hydrochloride salt.
Description 98% by HPLC
Target Proteins Proteasome
Molecular Formula C33H44N10O7·2HCl
Formula Weight 765.7g/mol
Solubility DMSO, MeOH, or Acetic Acid
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.