Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protein Arginine N-Methyltransferase Inhibitor, AMI-1

Catalog No. 5392095MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-920-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-920-95MG Supplier MilliporeSigma™ Supplier No. 5392095MG
Add to Cart
Add to Cart

The Protein Arginine N-Methyltransferase Inhibitor, AMI-1 controls the biological activity of Protein Arginine N-Methyltransferase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 95% by HPLC
Target Proteins Protein arginine N-methyltransferases (PRMTs)
Molecular Formula C21H12N2Na4O9S2
Formula Weight 592.4g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.