Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protein Lysine Methyltransferase Inhibitor, UNC0321

Catalog No. 4764965MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-649-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-649-65MG Supplier MilliporeSigma™ Supplier No. 4764965MG
Add to Cart
Add to Cart

A cell-permeable, quinazoline analog that potently and selectively inhibits PKMT G9a (IC50 = 6nM and 9nM in two biochemical assays for CLOT and ECSD, respectively, and Ki = 63 pM, which is approximately 250-fold more potent than a closely-related analog, BIX0129

A cell-permeable, quinazoline analog that potently and selectively inhibits PKMT G9a (IC50 = 6nM and 9nM in two biochemical assays for CLOT and ECSD, respectively, and Ki = 63 pM, which is approximately 250-fold more potent than a closely-related analog, BIX01294 (Cat. No. 382190)). It inhibits GLP with reduced potency (15nM) and is found to be inactive (IC50 > 40μM) toward other protein lysine and arginine methyltransferases, such as SET7/9 (aH3K4 PKMT), SET8/PreSET7 (aH4K20 PKMT), and PRMT3, as well as JMJD2E (> 1000-fold selectivity) in ECSD enzymatic assays. It also inhibits H3K9me2 accumulation (IC50 = 11,000nM) in MDA-MB-231 cells, in comparison to BIX01294 (IC50 = 500nM).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Tan
Description >98% by HPLC
Molecular Formula C27H45N7O3
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.