Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protein Synthesis Initiation Inhibitor, NSC 119889

Catalog No. 53969025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-969-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-969-025MG Supplier MilliporeSigma™ Supplier No. 53969025MG
Add to Cart
Add to Cart

The Protein Synthesis Initiation Inhibitor, NSC 119889 blocks 5'-mediated/cap-dependent initiation of protein synthesis. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Light Brown
Description 95% by HPLC
Target Proteins human arginine methyl transferase
Molecular Formula C20H8Br4O5
Formula Weight 647.9g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.