Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PSA Substrate, Fluorogenic

Catalog No. 5395821MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-958-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-958-21MG Supplier MilliporeSigma™ Supplier No. 5395821MG

An excellent fluorogenic substrate for assaying the proteolytic activity of prostate-specific antigen (PSA).

Peptide Sequence: Mu-His-Ser-Ser-Lys-Leu-Gln-AFC, 2TFA (Mu = morpholinoureidyl)
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 90% by HPLC
Target Proteins Fluorogenic substrate for assaying the proteolytic activity of prostate-specific antigen (PSA)
Molecular Formula C44H60N12O13F3·2HOAc
Formula Weight 1142g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.