Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PTP LYP Inhibitor

Catalog No. 5402175MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-021-75MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 54-021-75MG Supplier MilliporeSigma™ Supplier No. 5402175MG
Add to Cart
Add to Cart

The PTP LYP Inhibitor controls the biological activity of PTP LYP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

A cell-permeable, Au(I)-phosphine complex that acts as selective inhibitor of lymphoid tyrosine phosphatase (LYP) (IC50= 1.5μM) and has a ∽10-fold selectivity for LYP over PTP-PEST, HePTP, and CD45 in both JTAg cells and primary mouse thymocytes.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Molecular Formula C17H14AuClNP
Formula Weight 495.69g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.