Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pyridoxal, Hydrochloride

Catalog No. 54506025GM Shop All MilliporeSigma Products
Change view
Click to view available options
Pyridoxal, Hydrochloride; Qty: 25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
54-506-025GM Pyridoxal, Hydrochloride; Qty: 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 54-506-025GM Supplier MilliporeSigma™ Supplier No. 54506025GM
Add to Cart
Add to Cart

Helps in scavenging of free radical species and carbonyl species formed in sugar and lipid degradation

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Additive Pyridoxal, Hydrochloride
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.