Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pyridoxamine, Hydrochloride

Catalog No. 5450685GM Shop All MilliporeSigma Products
Change view
Click to view available options
Pyridoxamine, Hydrochloride; Qty: 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
54-506-85GM Pyridoxamine, Hydrochloride; Qty: 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 54-506-85GM Supplier MilliporeSigma™ Supplier No. 5450685GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Helps in scavenging of free radical species and carbonyl species formed in sugar and lipid degradation

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Additive Pyridoxamine, Hydrochloride
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.