Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ RAD51-Stimulatory Compound-1, RS-1

Catalog No. 55351010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-351-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-351-010MG Supplier MilliporeSigma™ Supplier No. 55351010MG
Add to Cart
Add to Cart

A cell-permeable sulfonamido-benzamide-based allosteric regulator that stimulates DNA binding and recombinase activities of hRAD51 by locking hRAD51 in an active conformation without affecting its active site ATP hydrolysis.

A cell-permeable sulfonamido-benzamide-based allosteric regulator that stimulates DNA binding and recombinase activities of hRAD51 by locking hRAD51 in an active conformation without affecting its active site ATP hydrolysis. Although RS-1 enhances hRAD51 filament formation on ssDNA with or without the cofactor NTP, active filaments and recombinase activity are only induced in the presence of ATP or AMP-PNP, but not with ADP or no cofactors. Shown to promote resistance of primary human neonatal dermal fibroblasts to Cisplatin- (Cat. No. 232120) induced death in a dose-dependent manner. RS-1 is inactive toward related DNA strand exchange proteins scRAD5 and scDMC1 of yeast origin or E. coli RedA.This HDR (homology-directed repair) enhancer, is shown to significantly increase both Cas9 and TALEN-mediated knock-in efficiencies.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Beige
Description 95% by HPLC
CAS 312756-74-4
Molecular Formula C20H16Br2N2O3S
Formula Weight 524.2g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.