Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ RAF Kinase Inhibitor VII

Catalog No. 5530185MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-301-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-301-85MG Supplier MilliporeSigma™ Supplier No. 5530185MG
Add to Cart
Add to Cart

A cell-permeable imidazopyridinyl-urea compound that acts as a potent inhibitor against B-RafV600E activity (IC50 = 1nM) by targeting its ATP-binding pocket and the adjacent lipophilic allosteric site accessible only when the enzyme is in its inactive DFG-out confor

A cell-permeable imidazopyridinyl-urea compound that acts as a potent inhibitor against B-RafV600E activity (IC50 = 1nM) by targeting its ATP-binding pocket and the adjacent lipophilic allosteric site accessible only when the enzyme is in its inactive DFG-out conformation. Reported to effectively inhibit RafV600E-dependent ERK phosphorylation (IC50 = 610nM) and growth of human melanoma WM266.4 cells (GI50 = 470nM).
TRUSTED_SUSTAINABILITY

Specifications

Color Caramel
Molecular Formula C21H15ClF3N5O3S
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.