Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Rapid Cell Proliferation Kit

Catalog No. QIA1271KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Each/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QIA1271KIT 1 Each/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QIA1271KIT Supplier MilliporeSigma™ Supplier No. QIA1271KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Actively drive reagents through membrane.

  • Vacuum-driven technology
  • Detection method: Chemiluminescence Fluorescence
  • Built-in flow distributor
  • Actively drive reagents through membrane
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Format Kit
Includes Calibration Standard, Positive Control Calpain 1, Substrate, Activation Buffer, Inhibition Buffer, Reduction Agent, Assay Buffer, Cell Lysis Buffer, 96-Well Plate, Plate Sealer, and user protocol.
Product Type Cell Proliferation Kit
Quantity 1 Each/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.