Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Reboxetine Mesylate

Catalog No. 5041200001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-412-00001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-412-00001 Supplier MilliporeSigma™ Supplier No. 5041200001
Add to Cart
Add to Cart

A potent and selective inhibitor of noradrenalin uptake (Ki values ar

A potent and selective inhibitor of noradrenalin uptake (Ki values are 1.1, 129 and >10000nM for rat NET, SERT and DAT, respectively). Displays >1000-fold selectivity over α-adrenoceptors, 5-HT, dopamine and muscarinic ACh receptors. Orally active antidepressant.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins Noradrenalin transporter
Molecular Formula C19H23NO3·CH3SO3H
Formula Weight 409.5g/mol
Solubility H2O
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.