Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Remodelin

Catalog No. 5310660001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-106-60001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-106-60001 Supplier MilliporeSigma™ Supplier No. 5310660001
Add to Cart
Add to Cart

A cell-permeable, potent, and stable analog of CPTH2 that restores nuclear shape and chromatin compactin in progeric fibroblasts.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 94% by HPLC
Target Proteins NAT10
Molecular Formula C15H14N4S·HBr
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.