Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Reserpine

Catalog No. 5061300001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-613-00001 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-613-00001 Supplier MilliporeSigma™ Supplier No. 5061300001
Add to Cart
Add to Cart

Binds the vesicular monoamine transporter (VMAT2) and acts as an irreversible transport inhibitor of biogenic amines.

Binds the vesicular monoamine transporter (VMAT2) and acts as an irreversible transport inhibitor of biogenic amines. Used as a long-lasting bioamine depleter. Prevents bioamine transport into synaptic vesicles and chromaffin granules. Used as antihypertensive and antipsychotic drugs.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins VMAT2
Molecular Formula C33H40N2O9
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.