Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Retro-2

Catalog No. 55471525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-471-525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-471-525MG Supplier MilliporeSigma™ Supplier No. 55471525MG
Add to Cart
Add to Cart

A cell-permeable tricyclic imine compound that inhibits the endosome-to-Golgi retrograde transport, but not cellular uptake, of CtxB, StxB without affecting cellular uptake/trafficking of Tf or EGF.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 95% by HPLC
Molecular Formula C19H16N2OS
Formula Weight 320.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.