Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Retromer Chaperone, R55

Catalog No. 5310840001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-108-40001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-108-40001 Supplier MilliporeSigma™ Supplier No. 5310840001
Add to Cart
Add to Cart

A cell-permeable pharmacological chaperone to increase neuronal retromer levels. Improves Vps35-Vps29-Vps26 retromer complex stability.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 92% by HPLC
Target Proteins vacuolar protein sorting (Vps) proteins
Molecular Formula C8H12N4S3·HCl
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.