Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ REV-ERB Agonist II, SR9009

Catalog No. 55472625MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-472-625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-472-625MG Supplier MilliporeSigma™ Supplier No. 55472625MG
Add to Cart
Add to Cart

The REV-ERB Agonist II, SR9009 controls the biological activity of REV-ERB. This small molecule/inhibitor is primarily used for Biochemicals applications.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Brown
Description 96% by HPLC
Target Proteins REV-ERB-α/β
Formula Weight 437.9g/mol
Solubility DMSO
Content And Storage Protect from light
Form Liquid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.