Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Rho Inhibitor II, Y16

Catalog No. 5040430001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-404-30001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-404-30001 Supplier MilliporeSigma™ Supplier No. 5040430001
Add to Cart
Add to Cart

A cell-permeable pyrazolidinedione compound that is shown to target RhoGEF DH-PH domain junction with high affinity (Kd = 65nM) and effetively prevent RhoGEFs LARG, p115, and PDZ from interacting with RhoA, while displaying little potency against DBL-RhoA, LBC-RhoA, intersectin-Cdc42, or TrioN-Cdc42 interact

A cell-permeable pyrazolidinedione compound that is shown to target RhoGEF DH-PH domain junction with high affinity (Kd = 65nM) and effetively prevent RhoGEFs LARG, p115, and PDZ from interacting with RhoA, while displaying little potency against DBL-RhoA, LBC-RhoA, intersectin-Cdc42, or TrioN-Cdc42 interaction. Shown to completely prevent serum-induced activation of cellular RhoA, but not Cdc42 or Rac1, in NIH-3T3 cultures (10μM) and RhoA downstream signaling events. Greatly synergizes with Rho GEF-binding domain blocker Rhosin (Cat. No. 555460) in blocking RhoA-LARG interaction and in preventing cellular RhoA activation (both drug at 5μM).
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 95% by HPLC
Target Proteins RhoGEF
Molecular Formula C24H20N2O3
Formula Weight 384.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.