Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Riboflavin

Catalog No. 55568210GM Shop All MilliporeSigma Products
Change view
Click to view available options
Riboflavin; Qty: 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
55-568-210GM Riboflavin; Qty: 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-568-210GM Supplier MilliporeSigma™ Supplier No. 55568210GM
Add to Cart
Add to Cart

Involved in synthesis of flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD)

  • Water soluble vitamin
  • Provides protection against oxidative damage
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Additive Riboflavin
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.