Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ RNA Polymerase I Inhibitor II, BMH-21

Catalog No. 5099110001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-991-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-991-10001 Supplier MilliporeSigma™ Supplier No. 5099110001
Add to Cart
Add to Cart

A cell-permeable compound that blocks the growth and viability of a variety of cancer cell lines (GI50 = 160 nM in NCI-60 cells), but does not affect normal cells.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Orange/Yellow
Description 98% by HPLC
Target Proteins rDNA
Molecular Formula C21H20N4O2
Solubility DMSO or 5% Aqueous Acetic Acid
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.