Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ RNA Polymerase III Inhibitor

Catalog No. 55740310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-740-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-740-310MG Supplier MilliporeSigma™ Supplier No. 55740310MG
Add to Cart
Add to Cart

RNA Polymerase III Inhibitor, CAS 577784-91-9, is a cell-permeable inhibitor of RNA Polymerase III (IC50 = 27 and 32 μM for human and S. cerevisiae RNA Pol III, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins activity against RNA Polymerase III
CAS 577784-91-9
Molecular Formula C19H15Cl2N3O2S2
Formula Weight 452.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.