Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ro-31-7549, Monohydrate

Catalog No. 5575081MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-750-81MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-750-81MG Supplier MilliporeSigma™ Supplier No. 5575081MG
Add to Cart
Add to Cart

A cell permeable, reversible, selective protein kinase C (PKC) inhibitor (IC50 = 158 nM for rat brain PKC) that acts at the ATP binding site of PKC.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Red to Reddish Orange
Description 93% by HPLC
Target Proteins rat brain PKC
CAS 125313-65-7
Molecular Formula C24H22N4O2·C2H4O2·H2O
Formula Weight 476.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.