Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ro106-9920

Catalog No. 5575501MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-755-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-755-01MG Supplier MilliporeSigma™ Supplier No. 5575501MG
Add to Cart
Add to Cart

A cell-permeable tetrazolopyridazine-phenylsulfoxide compound that displays anti-inflammatory properties.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins IκBαee ubiquitination
Molecular Formula C10H7N5OS
Formula Weight 245.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.