Learn More
MilliporeSigma™ Calbiochem™ RORγt Inverse Agonist II, GSK805
Catalog No. 5313690001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
53-136-90001 10 mg
1 options
Catalog No. 53-136-90001 Supplier MilliporeSigma™ Supplier No. 5313690001
Add to Cart
Add to Cart
A cell-permeable, potent, non-toxic, inverse agonist of RORγt. Directly and reversibly interacts with the ligand binding domain of RORγt to block its transcriptional effects.
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg