Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Rosiglitazone

Catalog No. 55736610MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-736-610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-736-610MG Supplier MilliporeSigma™ Supplier No. 55736610MG
Add to Cart
Add to Cart

A thiazolidinedione compound that acts as an anti-diabetic agent and serves as a potent and selective agonist of peroxisome proliferator-activated receptor-γ (PPARγ) (Kd ∽40 nM) in fat cells.

A thiazolidinedione compound that acts as an anti-diabetic agent and serves as a potent and selective agonist of peroxisome proliferator-activated receptor-g (PPARg) (Kd ∽40 nM) in fat cells. Shown to reduce fatty acid uptake and ameliorate lipid metabolism and insulin resistance in animal models of type II diabetes. Reported to activate both a1- and a2-containing AMPK complexes. Unlike troglitazone, it does not induce the activity of P4503A4. Significantly improves the differentiation of C3H10T1/2 stem cells into adipocytes. Shown to block estrogen synthesis by interfering with androgen binding to aromatase, but without affecting aromatase mRNA or protein expression.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
CAS 155141-29-0
Molecular Formula C18H19N3O3S·C4H4O4
Formula Weight 473.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.