Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ RuBi-Nicotine

Catalog No. 5052220001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-522-20001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-522-20001 Supplier MilliporeSigma™ Supplier No. 5052220001
Add to Cart
Add to Cart

A highly water soluble Ruthenium-bisbipyridine-caged nicotine that can be un-caged by blue and green light

A highly water soluble Ruthenium-bisbipyridine-caged nicotine that can be un-caged by blue and green light. Rapid nicotine release (< 20 ns) and fast biological effects in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Deep Orange
Description 98% by NMR
Target Proteins nAChR
Molecular Formula C40H44Cl2N8Ru
Formula Weight 808.81g/mol
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.