Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ RubR1 Activator, GF-15

Catalog No. 5004950001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-049-50001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-049-50001 Supplier MilliporeSigma™ Supplier No. 5004950001
Add to Cart
Add to Cart

A cell-permeable 2'-benzyloxy griseofulvin derivative that displays enhanced mammalian cancer-selective toxicity (IC50 ≤2, ≤4, ≤12, and ≥30μM against panels of leukemia, myeloma, solid tumors, and non-malignant cell lines, respectively), but little antifungal activity, by activating spindle-assembly-checkpoint without affecting microtubule kinetochore attachm

A cell-permeable 2'-benzyloxy griseofulvin derivative that displays enhanced mammalian cancer-selective toxicity (IC50 ≤2, ≤4, ≤12, and ≥30μM against panels of leukemia, myeloma, solid tumors, and non-malignant cell lines, respectively), but little antifungal activity, by activating spindle-assembly-checkpoint without affecting microtubule kinetochore attachment. Cancer cells are reported to undergo multipolar (MP) mitosis with normal or supernumerary centrosomes in the presence of GF-15, resulting in subsequent apoptotic cell death. Shown to retard HT29 colon cancer & OPM2 myeloma tumor expansion in mice (20mg/kg/day i.p.) in vivo . GF-15 half-life is expected to be least 48 h in cultures.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Molecular Formula C23H21ClO6
Formula Weight 428.9g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.