Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ryanodine, High Purity

Catalog No. 559276500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-927-6500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-927-6500UG Supplier MilliporeSigma™ Supplier No. 559276500UG
Add to Cart
Add to Cart

Naturally occurring alkaloid that blocks the release of Ca2+ from the sarcoplasmic reticulum.

Naturally occurring alkaloid that blocks the release of Ca2+ from the sarcoplasmic reticulum. Has many complex effects on Ca2+ transport and action in muscle.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Ca2+ transport and action in muscle
CAS 15662-33-6
Molecular Formula C25H35NO9
Formula Weight 493.6g/mol
Solubility MeOH or H2O
Content And Storage Protect from light
Form Solid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.