Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ S-Methyl-L-thiocitrulline, Dihydrochloride

Catalog No. 47280410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-280-410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-280-410MG Supplier MilliporeSigma™ Supplier No. 47280410MG
Add to Cart
Add to Cart

A cell-permeable inhibitor of nitric oxide synthase that exhibits about 17-fold greater selectivity for rat neuronal nitric oxide synthase (IC50 = 300nM) compared to the endothelial enzyme (IC50 = 5.4μM).

A cell-permeable inhibitor of nitric oxide synthase that exhibits about 17-fold greater selectivity for rat neuronal nitric oxide synthase (IC50 = 300nM) compared to the endothelial enzyme (IC50 = 5.4μM).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 90% by TLC
Target Proteins Rat neuronal nitric oxide synthase
CAS 209589-59-3
Molecular Formula C7H15N3O2S·2HCl
Formula Weight 278.2g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.