Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Salvinorin A

Catalog No. 5081590001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-815-90001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-815-90001 Supplier MilliporeSigma™ Supplier No. 5081590001
Add to Cart
Add to Cart

A highly potent and selective non-alkaloid positive allosteric modulator of κ-opioid receptors (Ki

A highly potent and selective non-alkaloid positive allosteric modulator of κ-opioid receptors (Ki = 4.3nM and 16nM for native and cloned κ-opioid receptors, respectively). Also acts as a D2DR partial agonist (Ki = 5-10nM). Used in studies of addition.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins k opioid receptor
Molecular Formula C23H28O8
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.