Learn More
MilliporeSigma™ Calbiochem™ SAM Methyltransferase Assay
Catalog No. CBA0961KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests/Kit
Catalog No. Quantity
CBA0961KIT 100 Tests/Kit
1 options
Catalog No. CBA0961KIT Supplier MilliporeSigma™ Supplier No. CBA0961KIT
Add to Cart
Add to Cart
Assays signal transduction for methytransferases
  • Continuous enzyme coupled assay
  • Monitors SAM dependent methyltransferases without use of radioactive labels or endpoint measurements
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 Tests/Kit
Content And Storage <-70°C