Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 202190, Hydrochloride

Catalog No. 5593931MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-939-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-939-31MG Supplier MilliporeSigma™ Supplier No. 5593931MG
Add to Cart
Add to Cart

A water-soluble form of the potent p38 MAP kinase inhibitor SB 202190.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Amber
Description 97% by HPLC
Target Proteins p38β
CAS 350228-36-3
Molecular Formula C20H14FN3O·HCl
Formula Weight 367.8g/mol
Solubility DMSO or slightly soluble in H2O (0.5 mg/ml)
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.