Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 206553 Hydrochloride

Catalog No. 5062720001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-627-20001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-627-20001 Supplier MilliporeSigma™ Supplier No. 5062720001
Add to Cart
Add to Cart

A selective 5-HT2C serotonin receptor inhibitor with lesser affinity for 5-HT2B

A selective 5-HT2C serotonin receptor inhibitor with lesser affinity for 5-HT2BR. Often used in conjunction with ketanserin to distinguish 5-HT2AR and 5-HT2CR activity. SB206553 has highest affinity for 5-HT2CR (Ki = 3.2nM) and less for 5-HT2BR (Ki = 5.5nM) and 5-HT2AR (Ki = 2,300nM).
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 99% by HPLC
Target Proteins 5-HT2C
Molecular Formula C17H16N4O·HCl
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.