Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 242084 Hydrochloride

Catalog No. 5064170001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-641-70001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-641-70001 Supplier MilliporeSigma™ Supplier No. 5064170001
Add to Cart
Add to Cart

A selective, brain-penetrant 5-HT2C receptor antagonist (pK

A selective, brain-penetrant 5-HT2C receptor antagonist (pKi 8.2-9.0 ), with some effects on the 5-HT2B receptor (pKi 6.8-7.0) and 5-HT2A receptor (pKi 6.1-6.8). SB 242084 has been shown to increase the effectiveness of the selective serotonin reuptake inhibitor (SSRI) class of antidepressants, and may also reduce their side effects. In animal studies, SB-242,084 produced stimulant-type activity and reinforcing effects, somewhat similar to but much weaker than cocaine or amphetamines. In vivo (ocular) IC50 values reported at 2.08μM.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins 5-HT2C
Molecular Formula C21H19ClN4O2·HCl
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.