Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB-269970 Hydrochloride

Catalog No. 5064150001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-641-50001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-641-50001 Supplier MilliporeSigma™ Supplier No. 5064150001
Add to Cart
Add to Cart

A selective 5-HT7 receptor antagon

A selective 5-HT7 receptor antagonist. Extremely potent in vivo and in vitro . Chronic or acute (even single dose) administration of SB269970 induces functional desensitization of the 5-HT7 receptor system, which precedes changes in the receptor density. This mechanism may be responsible for the rapid antidepressant-like effect of the 5-HT7 antagonist in animal models. SB-269970 is an analogue of SB-258719 and has been shown to have at least 100 fold selectivity versus all other 5-HT receptor subtypes except the human 5-HT5A receptor (pKi values are 8.9, 7.2 and 6.0 for 5-HT7A, 5-HT5A and 5-HT1B).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins 5-HT7
Molecular Formula C18H28N2O3S·HCl
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.