Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SB 290157

Catalog No. 55941010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-941-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-941-010MG Supplier MilliporeSigma™ Supplier No. 55941010MG
Add to Cart
Add to Cart

A selective, high affinity, competitive antagonist of the anaphylatoxin C3a receptor. Does not antagonize C5aR or other chemotactic GPCRs.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Description 95% by HPLC
Target Proteins C3a receptor
CAS 1140525-25-2
Molecular Formula C22H28N4O4·CF3CO2H
Formula Weight 526.5g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.