Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Scopolamine Hydrobromide

Catalog No. 5058180001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-581-80001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-581-80001 Supplier MilliporeSigma™ Supplier No. 5058180001
Add to Cart
Add to Cart

A competitive, non-selective antagonist for muscarinic acetylcholine receptors (mAChR)

A competitive, non-selective antagonist for muscarinic acetylcholine receptors (mAChR). Most effective at treating motion sickness. It is also a research tool for inducing memory impairment in vivo . Frequently used in researches related to learning, memory and Alzheimer′s dementia.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Molecular Formula C17H21NO4·HBr
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.