Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ γ-Secretase Inhibitor I

Catalog No. 5657501MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-575-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-575-01MG Supplier MilliporeSigma™ Supplier No. 5657501MG
Add to Cart
Add to Cart

The γ-Secretase Inhibitor I controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Z-Leu-Leu-Nle-CHO (Nle = Norleucine)
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 90% by HPLC
Target Proteins γ-secretase
Molecular Formula C26H41N3O5
Formula Weight 475.6g/mol
Solubility 50% Acetic Acid or DMSO
Content And Storage Hygroscopic
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.