Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ γ-Secretase Substrate, Fluorogenic

Catalog No. 5657641MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-576-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-576-41MG Supplier MilliporeSigma™ Supplier No. 5657641MG
Add to Cart
Add to Cart

An internally quenched fluorogenic peptide substrate containing the C-terminal β-APP (amyloid β-peptide precursor protein) amino acid sequence that is cleaved by γ-secretase.

  • Packaged Under Inert Gas
  • Peptide Sequence: NMA-Gly-Gly-Val-Val-Ile-Ala-Thr-Val-Lys(DNP)-D-Arg-D-Arg-D-Arg-NH2
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins γ-secretase
Molecular Formula C70H116N26O18
Formula Weight 1609.9g/mol
Solubility DMSO or 5% Acetic Acid
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.