Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sertraline Hydrochloride

Catalog No. 5061880001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-618-80001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-618-80001 Supplier MilliporeSigma™ Supplier No. 5061880001
Add to Cart
Add to Cart

A very potent uptake inhibitor highly selective for serotonin (Ki

A very potent uptake inhibitor highly selective for serotonin (Ki = 3.3nM). Clinically prescribed as an antidepressant drug. Often used in studying major depressive disorders.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins 5-HT uptake transporter
Molecular Formula C17H17Cl2N·HCl
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.