Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ sFRP-1 Inhibitor

Catalog No. 3443005MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-430-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-430-05MG Supplier MilliporeSigma™ Supplier No. 3443005MG
Add to Cart
Add to Cart

The sFRP-1 Inhibitor, also referenced under CAS 915754-88-0, controls the biological activity of sFRP-1. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description 95% by HPLC
CAS 915754-88-0
Molecular Formula C19H26N2O4S2
Formula Weight 410.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Oil
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.