Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Shikonin

Catalog No. 56585010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-585-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-585-010MG Supplier MilliporeSigma™ Supplier No. 56585010MG
Add to Cart
Add to Cart

A naphthoquinone found in the Chinese herb Shiunko.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description 98% by HPLC
Target Proteins TNF-α-induced and B-16 melanoma-induced angiogenesis
CAS 517-89-5
Molecular Formula C16H16O5
Formula Weight 288.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.