Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SHIP2 Inhibitor II, CPDA

Catalog No. 5083180001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-831-80001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-831-80001 Supplier MilliporeSigma™ Supplier No. 5083180001
Add to Cart
Add to Cart

A cell-permeable aminopyridine acetamide based compound that acts as an inhibitor of SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) that can overcome insulin resistance in 3T3-L1 adipocytes.

Enhances insulin-stimulated Akt phosphorylation (Thr308) in TNFa-treated 3T3-L1 adipocytes (∽10μM) and in primary cultured neurons from rat cerebral cortex. Improves glucose tolerance in db/db mice (∽300mg/kg b.i.d) without affecting their fasting glucose levels. Also reduces the expression levels of glucose-6-phosphatase and phosphoenolpyruvate carboxykinase.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins SHIP2
Molecular Formula C20H15ClF2N2O2
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.