Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SIRT1 Inhibitor IV, (S)-35

Catalog No. 566325500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-632-5500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-632-5500UG Supplier MilliporeSigma™ Supplier No. 566325500UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The SIRT1 Inhibitor IV, (S)-35, also referenced under CAS 848193-72-6, controls the biological activity of SIRT1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 96% by HPLC
CAS 848193-72-6
Molecular Formula C14H15ClN2O
Formula Weight 262.7g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.