Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sirt1 Inhibitor VII, Inauhzin

Catalog No. 56633210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-633-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-633-210MG Supplier MilliporeSigma™ Supplier No. 56633210MG
Add to Cart
Add to Cart

The Sirt1 Inhibitor VII, Inauhzin controls the biological activity of Sirt1. This small molecule/inhibitor is primarily used for Cell Structure applications.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Description 98% by HPLC
Target Proteins Sirt1
Molecular Formula C25H19N5OS2
Formula Weight 469.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.