Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SKF-525A, Hydrochloride

Catalog No. 5673001GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-730-01GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-730-01GM Supplier MilliporeSigma™ Supplier No. 5673001GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell permeable. Blocks glibenclamide-sensitive K+ channels. Inhibits neuronal nitric oxide synthase. Also inhibits hepatic drug metabolism by inhibiting the cytochrome P450 system.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins nNOS
CAS 62-68-0
Molecular Formula C23H31NO2·HCl
Formula Weight 390g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.