Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SKP2 E3 Ligase Inhibitor II, SMIP004

Catalog No. 5005170001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-051-70001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-051-70001 Supplier MilliporeSigma™ Supplier No. 5005170001
Add to Cart
Add to Cart

A cell-permeable phenylacetamide compound that is shown to upregulate p21Cip1/Waf1 and/or p27Kip1 in prostate cancer LNCaP, PC3, DU145, but not HeLa, cultures (40μM for 24 h) with a concomitant downregulation of the F-box E3 ubiquitin ligase Skp2 and a delayed G1-to-S cell cycle transition, displaying no apparent effect toward cell cycle progression or p21 and p27 levels in non-cancer human fibroblast

A cell-permeable phenylacetamide compound that is shown to upregulate p21Cip1/Waf1 and/or p27Kip1 in prostate cancer LNCaP, PC3, DU145, but not HeLa, cultures (40μM for 24 h) with a concomitant downregulation of the F-box E3 ubiquitin ligase Skp2 and a delayed G1-to-S cell cycle transition, displaying no apparent effect toward cell cycle progression or p21 and p27 levels in non-cancer human fibroblast IMR90. SMIP004 antiproliferative activity against LNCaP, PC3, DU145 soft agar colony formation is mainly cytostatic with cytotoxicity observed only in LNCaP cultures at high concentrations (IC50 = 40μM in 72 h), but not PC3 or DU145 even at concentrations as high as 80μM and for up to 96 h.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Molecular Formula C13H19NO
Formula Weight 205.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.