Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SKP2 E3 Ligase Inhibitor III

Catalog No. 5063050001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-630-50001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-630-50001 Supplier MilliporeSigma™ Supplier No. 5063050001
Add to Cart
Add to Cart

A cell-permeable benzothiazolylchromenone compound that disrupts Skp2-Skp1 interaction via direct Skp2 affinity binding and selectively prevents Skp2-, but not Fbw7- or β-TrCP-, Skp1-Cullin-1-F-box (SFC) complex-mediated substrates ubiquitination, effectively preventing Skp2 substrates activation (Akt) and degradation (p21 & p27) with concomitant down-regulation of cellular Skp2 protein level (10 to 20μM in HEK293T & PC3 cultures)

A cell-permeable benzothiazolylchromenone compound that disrupts Skp2-Skp1 interaction via direct Skp2 affinity binding and selectively prevents Skp2-, but not Fbw7- or β-TrCP-, Skp1-Cullin-1-F-box (SFC) complex-mediated substrates ubiquitination, effectively preventing Skp2 substrates activation (Akt) and degradation (p21 & p27) with concomitant down-regulation of cellular Skp2 protein level (10 to 20μM in HEK293T & PC3 cultures). Exhibits antiproliferation activity toward a panel of 8 cancer (IC50 from 1.22 to 10.5μM) and reduces ALDH+ cancer stem cell population in PC3 cultures (10 to 20μM). Exhibits good pharmacokinetic property and is efficacious in suppressing A549 and PC3 tumor growth in mice (40 to 80mg/kg/d i.p.) in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Description 98% by HPLC
Target Proteins Skp2
Molecular Formula C24H24N2O3S
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.