Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Smac-N7 Peptide, Cell-Permeable

Catalog No. 5673751MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-737-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-737-51MG Supplier MilliporeSigma™ Supplier No. 5673751MG
Add to Cart
Add to Cart

The Smac-N7 peptide is modified to be cell permeant by linking the lysine carboxyl terminal to the arginine of Antennapedia homeodomain 16-mer peptide via a proline linker.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Ala-Val-Pro-Ile-Ala-Gln-Lys-Pro-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Promotes procaspase-3 activation
Molecular Formula C142H232N44O29S
Formula Weight 3051.7g/mol
Solubility DMSO
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.